missing translation for 'onlineSavingsMsg'
Learn More

Imidazo[1,2-a]pyridine-6-carbaldehyde, ≥97%, Thermo Scientific™

Catalog Number p-3175388
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
116355-16-9
C8H6N2O
146.149
MFCD07772808
VDWYLKPRUHCLAW-UHFFFAOYSA-N
imidazo 1,2-a pyridine-6-carbaldehyde, imidazo 1,2-a pyridine-6-carboxaldehyde, 6-formylimidazo 1,2-a pyridine, pubchem23883, acmc-20a31e, 6-imidazo 1,2-a pyridinecarbaldehyde, 6-imidazo 1,2-a-pyridinecarbaldehyde, h-imidazo 1,2-a pyridine-6-carbaldehyde, 4-hydroimidazo 1,2-a pyridine-6-carbaldehyde
7164582
imidazo[1,2-a]pyridine-6-carbaldehyde
C1=CC2=NC=CN2C=C1C=O
This item is not returnable. View return policy
116355-16-9
C8H6N2O
146.149
MFCD07772808
VDWYLKPRUHCLAW-UHFFFAOYSA-N
imidazo 1,2-a pyridine-6-carbaldehyde, imidazo 1,2-a pyridine-6-carboxaldehyde, 6-formylimidazo 1,2-a pyridine, pubchem23883, acmc-20a31e, 6-imidazo 1,2-a pyridinecarbaldehyde, 6-imidazo 1,2-a-pyridinecarbaldehyde, h-imidazo 1,2-a pyridine-6-carbaldehyde, 4-hydroimidazo 1,2-a pyridine-6-carbaldehyde
7164582
imidazo[1,2-a]pyridine-6-carbaldehyde
C1=CC2=NC=CN2C=C1C=O
CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 146.15
Percent Purity 95%
Chemical Name or Material Imidazo[1, 2-a]pyridine-6-carbaldehyde
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'