Learn More
Thermo Scientific Maybridge Imidazo[1,2-a]pyridin-6-ylmethylamine, 1g, 97%
Brand: Thermo Scientific Maybridge CC33113DA
Imidazo[1,2-a]pyridin-6-ylmethylamine, 1g, 97%
Code : 31
Additional Details : CAS Number : 132213-03-7 Weight : 0.05100kg Transport : UN number : 3259 Chem class : 8 Pack group : III