Learn More
Imetit dihydrobromide, Tocris Bioscience™
Standard H3 and H4 agonist (H3 > H4)
Brand: Tocris (Bio-Techne) 0729/10
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity | 10mg |
---|
Description
An extremely potent, high affinity agonist at H3 and H4 receptors (Ki values are 0.3 and 2.7 nM respectively). Induces shape change in eosinophils with an EC50 of 25 nM. Centrally active following systemic administration. Also available as part of the Histamine H3 Receptor Tocriset™ .Chemical Identifiers
32385-58-3 | |
332.058 | |
imetit dihydrobromide, imetit hydrobromide, s-2-imidazol-4-yl ethyl isothiourea, s-2-1h-imidazol-4-yl ethyl isothiourea dihydrobromide, 2-1h-imidazol-4-yl ethyl carbamimidothioate dihydrobromide, s-2-imidazol-4-yl ethyl isothiourea dihydrobromide, imetit dihydrobromide hplc, s-2-imidazol-4-yl ethyl isothiourea 2hbr, s-2-imidazol-4-yl ethyl isothiourea dihydrochloride, 2-2-1h-imidazol-5-yl ethyl isothiourea dihydrobromide | |
CHEBI:64151 | |
C1=C(NC=N1)CCSC(=N)N.Br.Br |
C6H12Br2N4S | |
DOBOYMKCRRLTRF-UHFFFAOYSA-N | |
11957573 | |
2-(1H-imidazol-5-yl)ethyl carbamimidothioate;dihydrobromide |
Specifications
10mg | |
32385-58-3 | |
imetit dihydrobromide, imetit hydrobromide, s-2-imidazol-4-yl ethyl isothiourea, s-2-1h-imidazol-4-yl ethyl isothiourea dihydrobromide, 2-1h-imidazol-4-yl ethyl carbamimidothioate dihydrobromide, s-2-imidazol-4-yl ethyl isothiourea dihydrobromide, imetit dihydrobromide hplc, s-2-imidazol-4-yl ethyl isothiourea 2hbr, s-2-imidazol-4-yl ethyl isothiourea dihydrochloride, 2-2-1h-imidazol-5-yl ethyl isothiourea dihydrobromide | |
C1=C(NC=N1)CCSC(=N)N.Br.Br | |
332.058 | |
CHEBI:64151 | |
>98% |
Imetit dihydrobromide | |
C6H12Br2N4S | |
DOBOYMKCRRLTRF-UHFFFAOYSA-N | |
2-(1H-imidazol-5-yl)ethyl carbamimidothioate;dihydrobromide | |
11957573 | |
332.06 |