missing translation for 'onlineSavingsMsg'
Learn More

Imetit dihydrobromide, Tocris Bioscience™

Standard H3 and H4 agonist (H3 > H4)

£124.95 - £448.35

Chemical Identifiers

CAS 32385-58-3
Molecular Formula C6H12Br2N4S
Molecular Weight (g/mol) 332.058
InChI Key DOBOYMKCRRLTRF-UHFFFAOYSA-N
Synonym imetit dihydrobromide, imetit hydrobromide, s-2-imidazol-4-yl ethyl isothiourea, s-2-1h-imidazol-4-yl ethyl isothiourea dihydrobromide, 2-1h-imidazol-4-yl ethyl carbamimidothioate dihydrobromide, s-2-imidazol-4-yl ethyl isothiourea dihydrobromide, imetit dihydrobromide hplc, s-2-imidazol-4-yl ethyl isothiourea 2hbr, s-2-imidazol-4-yl ethyl isothiourea dihydrochloride, 2-2-1h-imidazol-5-yl ethyl isothiourea dihydrobromide
PubChem CID 11957573
ChEBI CHEBI:64151
IUPAC Name 2-(1H-imidazol-5-yl)ethyl carbamimidothioate;dihydrobromide
SMILES C1=C(NC=N1)CCSC(=N)N.Br.Br
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
13437923 View Documents Tocris (Bio-Techne)
0729/10
10mg £124.95
10mg View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 18-12-2019
Please to purchase this item. Need a web account? Register with us today!
13447923 View Documents Tocris (Bio-Techne)
0729/50
50mg £448.35
50mg View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 18-12-2019
Please to purchase this item. Need a web account? Register with us today!
Description

Description

An extremely potent, high affinity agonist at H3 and H4 receptors (Ki values are 0.3 and 2.7 nM respectively). Induces shape change in eosinophils with an EC50 of 25 nM. Centrally active following systemic administration. Also available as part of the Histamine H3 Receptor Tocriset™ .
Specifications

Chemical Identifiers

32385-58-3
332.058
imetit dihydrobromide, imetit hydrobromide, s-2-imidazol-4-yl ethyl isothiourea, s-2-1h-imidazol-4-yl ethyl isothiourea dihydrobromide, 2-1h-imidazol-4-yl ethyl carbamimidothioate dihydrobromide, s-2-imidazol-4-yl ethyl isothiourea dihydrobromide, imetit dihydrobromide hplc, s-2-imidazol-4-yl ethyl isothiourea 2hbr, s-2-imidazol-4-yl ethyl isothiourea dihydrochloride, 2-2-1h-imidazol-5-yl ethyl isothiourea dihydrobromide
CHEBI:64151
C1=C(NC=N1)CCSC(=N)N.Br.Br
C6H12Br2N4S
DOBOYMKCRRLTRF-UHFFFAOYSA-N
11957573
2-(1H-imidazol-5-yl)ethyl carbamimidothioate;dihydrobromide

Specifications

10mg
32385-58-3
imetit dihydrobromide, imetit hydrobromide, s-2-imidazol-4-yl ethyl isothiourea, s-2-1h-imidazol-4-yl ethyl isothiourea dihydrobromide, 2-1h-imidazol-4-yl ethyl carbamimidothioate dihydrobromide, s-2-imidazol-4-yl ethyl isothiourea dihydrobromide, imetit dihydrobromide hplc, s-2-imidazol-4-yl ethyl isothiourea 2hbr, s-2-imidazol-4-yl ethyl isothiourea dihydrochloride, 2-2-1h-imidazol-5-yl ethyl isothiourea dihydrobromide
C1=C(NC=N1)CCSC(=N)N.Br.Br
332.058
CHEBI:64151
>98%
Imetit dihydrobromide
C6H12Br2N4S
DOBOYMKCRRLTRF-UHFFFAOYSA-N
2-(1H-imidazol-5-yl)ethyl carbamimidothioate;dihydrobromide
11957573
332.06