Learn More
Imetit dihydrobromide, Tocris Bioscience™
Standard H3 and H4 agonist (H3 > H4)
Brand:  Tocris (Bio-Techne) 0729/50
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 50mg
 View more versions of this product
Product Code. 13447923
  • £448.35 / 50mg
Estimated Shipment date
from Supplier 20-11-2019
Description
Description
An extremely potent, high affinity agonist at H3 and H4 receptors (Ki values are 0.3 and 2.7 nM respectively). Induces shape change in eosinophils with an EC50 of 25 nM. Centrally active following systemic administration. Also available as part of the Histamine H3 Receptor Tocriset™ .
Specifications
Chemical Identifiers
32385-58-3
332.058
imetit dihydrobromide, imetit hydrobromide, s-2-imidazol-4-yl ethyl isothiourea, s-2-1h-imidazol-4-yl ethyl isothiourea dihydrobromide, 2-1h-imidazol-4-yl ethyl carbamimidothioate dihydrobromide, s-2-imidazol-4-yl ethyl isothiourea dihydrobromide, imetit dihydrobromide hplc, s-2-imidazol-4-yl ethyl isothiourea 2hbr, s-2-imidazol-4-yl ethyl isothiourea dihydrochloride, 2-2-1h-imidazol-5-yl ethyl isothiourea dihydrobromide
CHEBI:64151
C1=C(NC=N1)CCSC(=N)N.Br.Br
C6H12Br2N4S
DOBOYMKCRRLTRF-UHFFFAOYSA-N
11957573
2-(1H-imidazol-5-yl)ethyl carbamimidothioate;dihydrobromide
Specifications
50mg
32385-58-3
imetit dihydrobromide, imetit hydrobromide, s-2-imidazol-4-yl ethyl isothiourea, s-2-1h-imidazol-4-yl ethyl isothiourea dihydrobromide, 2-1h-imidazol-4-yl ethyl carbamimidothioate dihydrobromide, s-2-imidazol-4-yl ethyl isothiourea dihydrobromide, imetit dihydrobromide hplc, s-2-imidazol-4-yl ethyl isothiourea 2hbr, s-2-imidazol-4-yl ethyl isothiourea dihydrochloride, 2-2-1h-imidazol-5-yl ethyl isothiourea dihydrobromide
C1=C(NC=N1)CCSC(=N)N.Br.Br
332.058
CHEBI:64151
>98%
Imetit dihydrobromide
C6H12Br2N4S
DOBOYMKCRRLTRF-UHFFFAOYSA-N
2-(1H-imidazol-5-yl)ethyl carbamimidothioate;dihydrobromide
11957573
332.06