Learn More
IMD 0354, Tocris Bioscience™
Inhibitor of IKKβ
Brand:  Tocris (Bio-Techne) 261110
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 10mg
 View more versions of this product
Product Code. 13441574
  • £155.76 / 10mg
Estimated Shipment
28-06-2019
Description
Description
Inhibitor of IκB kinase-β (IKKβ) that blocks NF-κB nuclear translocation. Attenuates myocardial ischemia/reperfusion injury by decreasing expression of adhesion molecules ICAM-1 and P-selectin and inhibiting cytokine and chemokine production in cardiomyocytes. Induces G0/G1 cell cycle arrest and apoptosis in HMC-1 and breast cancer cells.
Specifications
Chemical Identifiers
978-62-1
383.674
ikk-2 inhibitor v, n-3,5-bis trifluoromethyl phenyl-5-chloro-2-hydroxybenzamide, n-3,5-bis trifluoromethyl phenyl-5-chloro-2-hydroxybenzamide, n-3,5-bis-trifluoromethylphenyl-5-chloro-2-hydroxybenzamide, benzamide, n-3,5-bis trifluoromethyl phenyl-5-chloro-2-hydroxy, benzamide, n-3,5-bis trifluoromethyl phenyl-5-chloro-2-hydroxy, kinome_1864, d0z5dm, 3',5'-salicyloxylidide, 5-chloro-alpha,alpha,alpha,alpha',alpha',alpha'-hexafluoro, imd hplc , solid
N-[3,5-bis(trifluoromethyl)phenyl]-5-chloro-2-hydroxybenzamide
C15H8ClF6NO2
CHILCFMQWMQVAL-UHFFFAOYSA-N
5081913
C1=CC(=C(C=C1Cl)C(=O)NC2=CC(=CC(=C2)C(F)(F)F)C(F)(F)F)O
Specifications
IMD 0354
978-62-1
ikk-2 inhibitor v, n-3,5-bis trifluoromethyl phenyl-5-chloro-2-hydroxybenzamide, n-3,5-bis trifluoromethyl phenyl-5-chloro-2-hydroxybenzamide, n-3,5-bis-trifluoromethylphenyl-5-chloro-2-hydroxybenzamide, benzamide, n-3,5-bis trifluoromethyl phenyl-5-chloro-2-hydroxy, benzamide, n-3,5-bis trifluoromethyl phenyl-5-chloro-2-hydroxy, kinome_1864, d0z5dm, 3',5'-salicyloxylidide, 5-chloro-alpha,alpha,alpha,alpha',alpha',alpha'-hexafluoro, imd hplc , solid
C1=CC(=C(C=C1Cl)C(=O)NC2=CC(=CC(=C2)C(F)(F)F)C(F)(F)F)O
383.674
383.67
10mg
C15H8ClF6NO2
CHILCFMQWMQVAL-UHFFFAOYSA-N
N-[3,5-bis(trifluoromethyl)phenyl]-5-chloro-2-hydroxybenzamide
5081913
>99%
Videos