missing translation for 'onlineSavingsMsg'
Learn More

iMAC2, Tocris Bioscience™

Suppressor of mitochondrial apoptosis

Brand:  RD Systems (Bio-Techne) 3794/10

Code : Z1

Additional Details : Weight : 0.00010kg

Quantity 10mg

 View more versions of this product

Product Code. 13433614

  • £174.93 / 10mg

Estimated Shipment date
from Supplier 30-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Inhibitor of mitochondrial apoptosis-induced channel (MAC) (IC50 = 28 nM). Reduces STS-induced apoptosis in FL5.12 cells by over 50%. Inhibits release of cytochrome c by Bid-induced Bax activation (IC50 = 0.68 μM).
Specifications

Chemical Identifiers

335166-00-2
542.112
imac2, 3,6-dibromo-9-2-fluoro-3-1-piperazinyl propyl-9h-carbazole dihydrochloride, 3,6-dibromo-9-2-fluoro-3-piperazin-1-yl propyl carbazole dihydrochloride
3,6-dibromo-9-(2-fluoro-3-piperazin-1-ylpropyl)carbazole;dihydrochloride
C19H22Br2Cl2FN3
JWCVUOLQOHIRML-UHFFFAOYSA-N
90488878
C1CN(CCN1)CC(CN2C3=C(C=C(C=C3)Br)C4=C2C=CC(=C4)Br)F.Cl.Cl

Specifications

10mg
335166-00-2
imac2, 3,6-dibromo-9-2-fluoro-3-1-piperazinyl propyl-9h-carbazole dihydrochloride, 3,6-dibromo-9-2-fluoro-3-piperazin-1-yl propyl carbazole dihydrochloride
C1CN(CCN1)CC(CN2C3=C(C=C(C=C3)Br)C4=C2C=CC(=C4)Br)F.Cl.Cl
542.112
542.11
iMAC2
C19H22Br2Cl2FN3
JWCVUOLQOHIRML-UHFFFAOYSA-N
3,6-dibromo-9-(2-fluoro-3-piperazin-1-ylpropyl)carbazole;dihydrochloride
90488878
>98%
Videos
Documents