missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ IL7 (Interleukin 7); Recombinant Human Protein
IL7 (Interleukin 7); Recombinant Human Protein
£3125.00 - £10980.00
Specifications
Molecular Weight (g/mol) | 17.5 kDa |
---|---|
Name | IL7 (Interleukin 7) CTS™ Recombinant Human Protein |
Purification Method | Gel-purified |
Storage Requirements | 2°C to 8°C |
Regulatory Status | For research use only. Not for use in diagnostic procedures |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10354640
|
Gibco™
CTP0071 |
100 μg |
£3125.00
Each |
Estimated Shipment: 07-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10171330
|
Gibco™
CTP0073 |
1 mg |
£10980.00
Each |
Estimated Shipment: 07-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Recombinant IL7 is a bioactive protein intended for use in cell culture applications. IL7 stimulates the proliferation of pre-B and pro-B cells. IL7 also supports the maturation of megakaryocytes. ED50 range = 0.2 to 1 ng⁄mL (Specific Activity: 5.0 x 10^6 to 1.0 x 10^6 units⁄mg), determined by the dose dependent proliferation of human PBMCs. Optimal concentration for individual application should be determined by a dose response assay.Specifications
17.5 kDa | |
Gel-purified | |
For research use only. Not for use in diagnostic procedures | |
IL-7 (Interleukin 7) | |
IL7 (Interleukin 7) CTS™ Recombinant Human Protein | |
E. coli | |
Interleukins |
IL7 (Interleukin 7) CTS™ Recombinant Human Protein | |
2°C to 8°C | |
1 Ea. | |
IL7 (Interleukin 7) CTS™ Recombinant Human Protein | |
Human | |
Cytokines and receptors | |
>95% by SDS-PAGE |
For Research Use Only. Not for use in diagnostic procedures.