missing translation for 'onlineSavingsMsg'
Learn More

Sino Biological™ IL1RAPL2 (IL1R9) Recombinant Human Protein, hIgG1-Fc Tag

Expressed from DNA sequence encoding extracellular domain (Met 1-Glu 356) of human IL1RAPL2 precursor fused to the Fc region of human IgG1

Brand:  Sino Biological™ 10156H02H5

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13462939

  • £57.87 / 5µg


Estimated Shipment
05-03-2019
Add to basket

Description

Description

  • Lyophilized in 100MM Glycine, 10MM NaCl , 50MM Tris , pH 7.5
Specifications

Specifications

Lyophilized
66kd
5μg
−20° to −70°C
<1 EU/μg
L1RAPL2
Cytokines and Receptors
IL1RAPL2 (IL1R9) Recombinant Human Protein
26280
≥90% by SDS-PAGE
Human
Recombinant
Interleukin-1 receptor accessory protein-like 2
Assay Standard
Mammalian
hIgG1-Fc Tag
Videos

For Research Use Only. Not for use in diagnostic procedures.