missing translation for 'onlineSavingsMsg'
Learn More

Sino Biological™ IL1RAP (IL1R3) Recombinant Human Protein, hIgG1-Fc.His Tag

Expressed from DNA sequence encoding extracellular domain (Met 1-Glu 359) of human IL1RAP fused to polyhistidine-tagged Fc region of human IgG1

Brand:  Sino Biological™ 10121H03H50

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13432939

  • £187.38 / 50µg


Estimated Shipment
14-05-2018
Add to basket

Description

Description

  • Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
Specifications

Specifications

Lyophilized
67.3kd
50μg
−20° to −70°C
<1 EU/μg
IL1RAP
Cytokines and Receptors
IL1RAP (IL1R3) Recombinant Human Protein
3556
≥90% by SDS-PAGE
Human
Truncated, Recombinant
Interleukin 1 receptor-like 1
Assay Standard
Mammalian
hIgG1-Fc.His Tag
Videos

For Research Use Only. Not for use in diagnostic procedures.