missing translation for 'onlineSavingsMsg'
Learn More

Sino Biological™ IL1R1 Recombinant Rat Protein, His Tag

Expressed from DNA sequence encoding rat IL1R1 extracellular domain (Met 1-Lys 352) fused to polyhistidine tag at C-terminus

Brand:  Sino Biological™ 80028R08H50

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15329526

  • £156.15 / 50µg


EU Stock 2
Add to basket

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Description

Description

  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
Specifications

Specifications

Analysis of protein stucture and used as immunogen, protein standard, or in cell biology research applications
25663
≥90% by SDS-PAGE
Rat
<1 EU/μg
IL1R1
Cytokines and Receptors, CD Proteins
IL1R1 Recombinant Rat Protein
Lyophilized
38.5kd
50μg
Truncated, Recombinant
Interleukin 1 Receptor Type 1
Assay Standard
Mammalian
His Tag
Videos