missing translation for 'onlineSavingsMsg'
Learn More

Sino Biological™ IL17RC Recombinant Human Protein, hIgG1-Fc Tag

Expressed from DNA sequence encoding extracellular domain of human IL17RC isoform 3 (Met 1- Ala 454) fused to Fc region of human IgG1 at C-terminus

Brand:  Sino Biological™ 11747H02H50

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15209115

  • £225.90 / 50µg


Estimated Shipment
15-06-2018
Add to basket

Description

Description

  • Formulation: 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween80
Specifications

Specifications

Lyophilized
≥90% by SDS-PAGE
Human
<1 EU/μg
IL17RC
Cytokines and Receptors
L17RC Recombinant Human Protein
75.3kd
50μg
Truncated, Recombinant
Interleukin 17 receptor C
Assay Standard
Mammalian
IgG1 Fc Tag
Videos

For Research Use Only. Not for use in diagnostic procedures.