missing translation for 'onlineSavingsMsg'
Learn More

IL-9 Monoclonal Antibody (MH9D1), eFluor™ 450, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 48-7098-42

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15509806

  • £248.00 / 100 tests
Estimated Shipment: 03-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Interleukin 9 (IL-9) is produced by activated T helper 2 (Th2) lymphocytes. IL-9 regulates many cell types involved in classical Th2-associated asthma responses, including B and T lymphocytes, mast cells, eosinophils, and epithelial cells. The mouse IL-9 cDNA encodes a 144 amino acid precursor protein with an 18 amino acid signal peptide that is cleaved to form the mature cysteine-rich protein with a molecular mass of 14 kDa. Native mouse IL-9 is heavily glycosylated. Recently, TGF beta has been reported to play an important role in the differentiation of IL-17-producing T helper cells and inducible regulatory T cells by reprogramming Th2 cells to secrete IL-9, thereby driving the differentiation of Th9 cells directly.

IL-9 Monoclonal antibody specifically detects IL-9 in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

IL-9
Monoclonal
5 μL/Test
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2
P15248
Il9
Affinity chromatography
RUO
3578
4° C, store in dark, DO NOT FREEZE!
Liquid
Flow Cytometry
MH9D1
eFluor 450
Il9
cytokine P40; homolog of mouse T cell and mast cell growth factor 40; HP40; IL9; IL-9; ILN; Interleukin; interleukin 9; Interleukin9; interleukin-9; P40; p40 cytokine; p40 T-cell and mast cell growth factor; T-cell growth factor P40
Mouse
100 Tests
Primary
Human
Antibody
IgG2a κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only.