missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-7R alpha/CD127 Goat anti-Mouse, Polyclonal, R&D Systems™
Goat Polyclonal Antibody has been used in 2 publications
POA
Specifications
Antigen | IL-7R alpha/CD127 |
---|---|
Immunogen | Mouse myeloma cell line NS0-derived recombinant mouse IL-7 R alpha/CD127 Glu21-Asp239 Accession # Q9R0C1 |
Gene ID (Entrez) | 3575 |
Gene Accession No. | Q9R0C1 |
Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative |
Description
IL-7R alpha/CD127 Polyclonal specifically detects IL-7R alpha/CD127 in Mouse samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.Specifications
IL-7R alpha/CD127 | |
3575 | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Primary | |
Detects mouse IL-7 R alpha in direct ELISAs and Western blots. In Western blots, less than 1% cross-reactivity with recombinant human IL-7 R alpha is observed. | |
Unconjugated | |
Western Blot, Flow Cytometry, CyTOF | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Mouse myeloma cell line NS0-derived recombinant mouse IL-7 R alpha/CD127 Glu21-Asp239 Accession # Q9R0C1 | |
Q9R0C1 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Goat | |
IgG | |
Affinity Purified | |
RUO |
For Research Use Only