missing translation for 'onlineSavingsMsg'
Learn More

IL-6, PerCP-eFluor™ 710, clone: MQ2-13A5, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 46-7069-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15549346

  • £91.74 / 25 tests


EU Stock 4
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: The MQ2-13A5 antibody reacts with human interleukin-6 (IL-6), a 21-28kDa cytokine secreted by a variety of cell types.Applications Reported: This MQ2-13A5 antibody has been reported for use in intracellular staining followed by flow cytometric analysis.Applications Tested: This MQ2-13A5 antibody has been pre-titrated and tested by intracellular staining and flow cytometric analysis of stimulated normal human peripheral blood cells. This can be used at 5 μL (0.125 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. PerCP-eFluor® 710 can be used in place of PE-Cy5, PE-Cy5.5 or PerCP-Cy5.5. PerCP-eFluor® 710 emits at 710 nm and is excited with the blue laser (488 nm). Please make sure that your instrument is capable of detecting this fluorochrome. For a filter configuration, we recommend using the 685 LP dichroic mirror and 710/40 band pass filter, however the 695/40 band pass filter is an acceptable alternative. Our testing indicates that PerCP-eFluor® 710 conjugated antibodies are stable when stained samples are exposed to freshly prepared 2% formaldehyde overnight at 4°C, but please evaluate for alternative fixation protocols.Excitation: 488 nm; Emission: 710 nm; Laser: Blue Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

IL-6
MQ2-13A5
PerCP-eFluor 710
Conjugated
Rat
25 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Flow Cytometry (Intracellular Staining)
5μL (0.125μg)/test
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Interleukin-6, IFNB2
IgG1
RUO
Primary
Human
Videos