Learn More
IL-6 Monoclonal Antibody (MQ2-13A5), PerCP-eFluor™ 710, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
Brand: Affymetrix eBioscience 46-7069-42
172.00 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.23750kg
Description
Description: The MQ2-13A5 antibody reacts with human interleukin-6 (IL-6), a 21-28 kDa cytokine secreted by a variety of cell types. Applications Reported: This MQ2-13A5 antibody has been reported for use in intracellular staining followed by flow cytometric analysis. Applications Tested: This MQ2-13A5 antibody has been pre-titrated and tested by intracellular staining and flow cytometric analysis of stimulated normal human peripheral blood cells. This can be used at 5 µL (0.125 µg) per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. PerCP-eFluor® 710 can be used in place of PE-Cy5, PE-Cy5.5 or PerCP-Cy5.5. PerCP-eFluor® 710 emits at 710 nm and is excited with the blue laser (488 nm). Please make sure that your instrument is capable of detecting this fluorochrome. For a filter configuration, we recommend using the 685 LP dichroic mirror and 710/40 band pass filter, however the 695/40 band pass filter is an acceptable alternative. Our testing indicates that PerCP-eFluor® 710 conjugated antibodies are stable when stained samples are exposed to freshly prepared 2% formaldehyde overnight at 4°C, but please evaluate for alternative fixation protocols. Excitation: 488 nm; Emission: 710 nm; Laser: Blue Laser. Filtration: 0.
2 μm post- manufacturing filtered. Interleukin 6 (IL-6) is a multifunctional 26 kD protein originally discovered in the medium of RNA-stimulated fibroblastoid cells. IL-6 appears to be directly involved in the responses that occur after infection and cellular injury, and it may prove to be important as IL-1 and TNF-a in regulating the acute phase response. IL-6 is reported to be produced by fibroblasts, activated T cells, activated monocytes or macrophages and endothelial cells. IL-6 acts upon a variety of cells including myeloid progenitor cells, T cells, B cells, and hepatocytes. In addition, IL-6 appears to interact with IL-2 in the proliferation of T lymphocytes. IL-6 potentiates the proliferative effect of IL-3 on multipotential hematopoietic progenitors. IL-6 plays a critical role in B-cell differentiation to plasma cells and is a potent growth factor for plasmacytoma and myeloma. IL-6 is a very useful culture supplement for the generation of a high number of antibody-producing hybridomas. Primarily produced at sites of acute and chronic inflammation, IL-6 is secreted into the serum and induces a transcriptional inflammatory response through interleukin 6 receptor, alpha. The functioning of IL-6 is implicated in a wide variety of inflammation-associated disease states including diabetes mellitus and systemic juvenile rheumatoid arthritis.Specifications
IL-6 | |
Monoclonal | |
5 μL/Test | |
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2 | |
P05231 | |
IL6 | |
Affinity chromatography | |
RUO | |
3569 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
MQ2-13A5 | |
PerCP-eFluor 710 | |
IL6 | |
Interleukin-6, BSF2, HGF, HSF, IFNB2, IL-6, B-cell stimulatory factor 2, BSF-2, Interferon beta-2, Hybridoma growth factor, CTL differentiation factor | |
Rat | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
For Research Use Only.