missing translation for 'onlineSavingsMsg'
Learn More

IL-6, PE-Cyanine7, clone: MQ2-13A5, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 25-7069-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15548626

  • £87.25 / 25 tests


Estimated Shipment date
from Supplier 20-02-2018
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: The MQ2-13A5 antibody reacts with human interleukin-6 (IL-6), a 21-28kDa cytokine secreted by a variety of cell types.Applications Reported: This MQ2-13A5 antibody has been reported for use in intracellular staining followed by flow cytometric analysis.Applications Tested: This MQ2-13A5 antibody has been pre-titrated and tested by intracellular staining and flow cytometric analysis of stimulated normal human peripheral blood cells using the Intracellular Fixation and Permeabilization Buffer Set (cat. 88-8824) and protocol. Please refer to Best Protocols: Protocol A: Two step protocol for (cytoplasmic)intracellular proteins located under the Resource Tab online. This can be used at 5 μL (0.06 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light.Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 μL of cell sample + 100 μL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically.Excitation: 488-561 nm; Emission: 775 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

IL-6
MQ2-13A5
PE-Cyanine7
Conjugated
Rat
25 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Monoclonal
Flow Cytometry (Intracellular Staining)
5μL (0.06μg)/test
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Interleukin-6
IgG1
RUO
Primary
Human
Videos