missing translation for 'onlineSavingsMsg'
Learn More

IL-6 Rat anti-Human, PE, Clone: MQ2-13A5, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 12-7069-41

Code : NEW

Additional Details : Weight : 0.23750kg

Product Code. 15577086

  • £65.50 / 25 tests

Estimated Shipment
04-11-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Interleukin 6 (IL-6) is a multifunctional 26 kD protein originally discovered in the medium of RNA-stimulated fibroblastoid cells. IL-6 appears to be directly involved in the responses that occur after infection and cellular injury, and it may prove to be important as IL-1 and TNF-a in regulating the acute phase response. IL-6 is reported to be produced by fibroblasts, activated T cells, activated monocytes or macrophages and endothelial cells. IL-6 acts upon a variety of cells including myeloid progenitor cells, T cells, B cells, and hepatocytes. In addition, IL-6 appears to interact with IL-2 in the proliferation of T lymphocytes. IL-6 potentiates the proliferative effect of IL-3 on multipotential hematopoietic progenitors. IL-6 plays a critical role in B-cell differentiation to plasma cells and is a potent growth factor for plasmacytoma and myeloma. IL-6 is a very useful culture supplement for the generation of a high number of antibody-producing hybridomas. Primarily produced at sites of acute and chronic inflammation, IL-6 is secreted into the serum and induces a transcriptional inflammatory response through interleukin 6 receptor, alpha. The functioning of IL-6 is implicated in a wide variety of inflammation-associated disease states including diabetes mellitus and systemic juvenile rheumatoid arthritis.
Specifications

Specifications

IL-6
Monoclonal
5 μL/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
P05231
Interleukin-6, BSF2, HGF, HSF, IFNB2, IL-6, B-cell stimulatory factor 2, BSF-2, Interferon beta-2, Hybridoma growth factor, CTL differentiation factor
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
MQ2-13A5
PE
IL6
Liquid
IL6
IgG1, kappa
25 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
3569
Videos
Documents

For Research Use Only.