missing translation for 'onlineSavingsMsg'
Learn More

IL-6, Functional Grade, clone: MQ2-13A5, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 16-7069-81

Code : Z2

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15218757

  • £62.18 / 50µg


Estimated Shipment
01-03-2018
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: The MQ2-13A5 antibody reacts with human interleukin-6 (IL-6), a 21-28kDa cytokine secreted by a variety of cell types.Applications Reported: This MQ2-13A5 antibody has been reported for use in cytokine neutralization, ELISPOT, and ELISA capture.Applications Tested: The MQ2-13A5 antibody has been tested for neutralizing IL-6 binding and biological activity.Endotoxin: Less than 0.001 ng/ug antibody as determined by the LAL assay.Storage and handling: Use in a sterile environment.Filtration: 0.2μm post-manufacturing filtered.Purity: Greater than 90%, as determined by SDS-PAGE.Endotoxin Level: Less than 0.001 ng/μg antibody, as determined by LAL assay.Aggregation: Less than 10%, as determined by HPLC.
Specifications

Specifications

IL-6
MQ2-13A5
Unconjugated
Purified
Rat
50μg
Store at 2-8°C.
Monoclonal
ELISPOT
1mg/mL
aqueous buffer, no sodium azide
Interleukin-6
IgG1
RUO
Primary
Human
Videos