missing translation for 'onlineSavingsMsg'
Learn More

IL-6, clone: MQ2-13A5, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 14-7069-85

Code : Z2

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15228007

  • £145.49 / 500µg


EU Stock 3
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: The MQ2-13A5 antibody reacts with human interleukin-6 (IL-6), a 21-28kDa cytokine secreted by a variety of cell types.Applications Reported: The MQ2-13A5 antibody has been reported for use in capture of human IL-6 by ELISA, intracellular staining for flow cytometric analysis, and for neutralizing IL-6 bioactivity.Applications Tested: The MQ2-13A5 antibody has been tested as the capture antibody in a sandwich ELISA for analysis of human Interleukin-6 (IL-6) in combination with the biotin MQ2-39C3 (13-7068) antibody for detection and recombinant human IL-6 (14-8069) as the standard. A suitable range of concentrations of this antibody for ELISA capture is 2-8 μg/mL. A standard curve consisting of doubling dilutions of the recombinant standard over the range of 1000 pg/mL - 8 pg/mL should be included in each ELISA plate.Purity: Greater than 90%, as determined by SDS-PAGE.Aggregation: Less than 10%, as determined by HPLC.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

IL-6
MQ2-13A5
Unconjugated
Purified
Rat
500μg
Store at 2-8°C.
Monoclonal
ELISPOT Capture Antibody
0.5mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Interleukin-6
IgG1
RUO
Primary
Human
Videos