Learn More
IL-6 Monoclonal Antibody (MQ2-13A5), Alexa Fluor™ 700, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
£242.00
Specifications
Antigen | IL-6 |
---|---|
Clone | MQ2-13A5 |
Concentration | 5 μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Description
Description: The MQ2-13A5 antibody reacts with human interleukin-6 (IL-6), a 21-28 kDa cytokine secreted by a variety of cell types. Applications Reported: This MQ2-13A5 antibody has been reported for use in intracellular staining followed by flow cytometric analysis. Applications Tested: This MQ2-13A5 antibody has been pre-titrated and tested by intracellular staining and flow cytometric analysis of stimulated normal human peripheral blood cells using the Intracellular Fixation & Permeabilization Buffer Set (cat. 88-8824) and protocol. Please refer to Best Protocols: Protocol A: Two step protocol for (cytoplasmic) intracellular proteins located under the Resources Tab online. This can be used at 5 μL (0.06 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Alexa Fluor™ 700 emits at 723 nm and can be excited with the red laser (633 nm). Most instruments will require a 685 LP mirror and 710/20 filter. Please make sure that your instrument is capable of detecting this fluorochrome. Excitation: 633-647 nm; Emission: 723 nm; Laser: Red Laser. Filtration: 0.2 μm post-manufacturing filtered.
IL-6 Monoclonal antibody specifically detects IL-6 in Human samples. It is validated for Flow Cytometry.Specifications
IL-6 | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
P05231 | |
IgG1 κ | |
Affinity chromatography | |
Antibody |
MQ2-13A5 | |
Flow Cytometry | |
Alexa Fluor 700 | |
Rat | |
Human | |
3569 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! |
For Research Use Only.