missing translation for 'onlineSavingsMsg'
Learn More

IL-5, Biotin, clone: JES1-5A10, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 13-7059-81

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15370020

  • £68.95 / 50µg


Estimated Shipment date
from Supplier 25-07-2018
Add to basket

Description

Description

Description: The JES1-5A10 antibody reacts with human interleukin-5 (IL-5), also known as BCDF-alpha and eosinophil differentiation factor (EDF). This 15-20kDa cytokine is secreted by T cells and exists a homodimer.Applications Reported: JES1-5A10 has been reported for use in detection of human IL-5 by ELISA.Applications Tested: The JES1-5A10 antibody has been tested as the detection antibody in a sandwich ELISA for analysis of human Interleukin-5 (IL-5) in combination with the purified TRFK5 (14-7052) antibody for capture and recombinant human IL-5 (14-8059) as the standard. A suitable range of concentrations of this antibody for ELISA detection is 0.5-2 μg/mL. A standard curve consisting of doubling dilutions of the recombinant standard over the range of 500 pg/mL - 4 pg/mL should be included in each ELISA plate.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

IL-5
Monoclonal
0.5mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
P05113
Interleukin-5, EDF, IL-5, TRF, T-cell replacing factor, TRF, Eosinophil differentiation factor, B cell differentiation factor I
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Human
ELISAþDot blot
JES1-5A10
Biotin
IL5
Conjugated
IL5
IgG2a, kappa
50μg
Store at 2-8°C. Do not freeze.
Primary
3567
Videos

For Research Use Only.