missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rat IL-5 Antibody, R&D Systems™
Mouse Monoclonal Antibody has been used in 1 publication
POA
Specifications
Antigen | IL-5 |
---|---|
Clone | 119905 |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Classification | Monoclonal |
Description
IL-5 Monoclonal specifically detects IL-5 in Rat samples. It is validated for Western Blot.Specifications
IL-5 | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
BCDF mu, B-cell differentiation factor I, BCGFII, EDF, Eo-CSF, Eosinophil differentiation factor, IL5, IL-5T-cell replacing factor, interleukin 5 (colony-stimulating factor, eosinophil), interleukin-5, TRF, TRFB cell differentiation factor I | |
IL5 | |
IgG2b | |
Protein A or G purified from ascites | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
119905 | |
Western Blot | |
Unconjugated | |
Mouse | |
Rat | |
Q08125 | |
3567 | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant rat IL-5 Met20-Val132 Accession # Q08125 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects rat IL-5 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human IL-5 and recombinant mouse IL-5 is observed. |
For Research Use Only