Learn More
IL-4 Monoclonal Antibody (11B11), PerCP-eFluor™ 710, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
£89.75 - £219.00
Specifications
Antigen | IL-4 |
---|---|
Clone | 11B11 |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15509346
|
Affymetrix eBioscience
46-7041-80 |
25 μg |
£89.75
25µg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15519346
|
Affymetrix eBioscience
46-7041-82 |
100 μg |
£219.00
100µg |
Estimated Shipment: 03-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Description: The 11B11 antibody reacts with mouse interleukin-4 (IL-4), a 14 kD cytokine. The 11B11 antibody is a neutralizing antibody. Mouse IL-4, also known as BCDF, BCGF-1 and BSF-1, is secreted by activated Th2 and natural killer T cells, and to a lesser extent by Th1 and mast cells. IL-4 is species-specific and promotes proliferation and differentiation of B cells. IL-4, in conjunction with other signals such as CD40, induces Ig isotype switching of B cells and the upregulation of surface MHC class II and CD23. Applications Reported: This 11B11 antibody has been reported for use in intracellular staining followed by flow cytometric analysis. Applications Tested: This 11B11 antibody has been tested by intracellular staining and flow cytometric analysis of Th2 polarized and stimulated mouse splenocytes. This can be used at less than or equal to 0.125 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. PerCP-eFluor™ 710 emits at 710 nm and is excited with the blue laser (488 nm); it can be used in place of PerCP-Cyanine5.5. We recommend using a 710/50 bandpass filter, however, the 695/40 bandpass filter is an acceptable alternative.
IL-4 Monoclonal antibody specifically detects IL-4 in Mouse samples. It is validated for Flow Cytometry.Specifications
IL-4 | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
B cell growth factor 1; B_cell stimulatory factor 1; B-cell growth factor 1; B-cell IgG differentiation factor; B-cell stimulatory factor 1; BCGF1; BCGF-1; binetrakin; BSF1; BSF-1; cytokine; H-IL-4; IGG1 induction factor; IL2; IL4; Il-4; Il4e12; ILN; Interleukin; interleukin 4; interleukin 4 variant 2; interleukin 4 variant IL-4delta2; interleukin 4 variant IL-4delta3; interleukin 4 variant IL-4int2A; interleukin 4 variant IL-4int3A; interleukin 4 variant IL-4int3B; Interleukin4; interleukin-4; interleukin-4 precursor; lymphocyte stimulatory factor 1; MGC79402; M-IL-4; Pitrakinra | |
Il4 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
Il4 |
11B11 | |
Flow Cytometry | |
PerCP-eFluor 710 | |
Rat | |
Mouse | |
P07750 | |
16189 | |
IgG1 κ | |
Affinity chromatography | |
Antibody |
For Research Use Only.