missing translation for 'onlineSavingsMsg'
Learn More

IL-4, PE, clone: 11B11, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 12-7041-82

Code : Z2

Additional Details : Weight : 0.00100kg

 View more versions of this product

Product Code. 12757327

  • £156.21 / 100µg


EU Stock 2
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: The 11B11 antibody reacts with mouse interleukin-4 (IL-4), a 14 kD cytokine. The 11B11 antibody is a neutralizing antibody. Mouse IL-4, also known as BCDF, BCGF-1 and BSF-1, is secreted by activated Th2 and natural killer T cells, and to a lesser extent by Th1 and mast cells. IL-4 is species-specific and promotes proliferation and differentiation of B cells. IL-4, in conjunction with other signals such as CD40, induces Ig isotype switching of B cells and the upregulation of surface MHC class II and CD23.Applications Reported: This 11B11 antibody has been reported for use in intracellular staining followed by flow cytometric analysis.Applications Tested: This 11B11 antibody is offered in 2 formats: -μg size: has been tested by intracellular staining of Th2 polarized and stimulated mouse splenocytes. This can be used at less than or equal to 0.125μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. - test size: has been pre-titrated and tested by intracellular staining of Th2 polarized and stimulated mouse splenocytes. This can be used at 5μL (0.06μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

IL-4
11B11
PE
Conjugated
Rat
100μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Flow Cytometry (Intracellular Staining)
0.2mg/mL; 5μL (0.06μg)/test
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Interleukin-4
IgG1
RUO
Primary
Murine
Videos