missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-4, FITC, clone: BVD6-24G2, eBioscience™
Rat Monoclonal Antibody
Brand: Affymetrix eBioscience 11-7042-41
Code : NEW
Additional Details : Weight : 0.23750kg
Description
Description: The BVD6-24G2 antibody reacts with mouse interleukin-4 (IL-4), a 14kDa cytokine secreted by Th2 cells.Applications Reported: BVD6-24G2 has been reported for use in intracellular flow cytometric analysis.Applications Tested: This BVD6-24G2 antibody has been pre-titrated and tested by intracellular staining and flow cytometric analysis of restimulated mouse splenocytes cultures. Refer to catalog number suffix on the vial for amount to use per test: -41 is 5μL (0.25μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser.Filtration: 0.2μm post-manufacturing filtered.Specifications
IL-4 | |
Monoclonal | |
5μl/Test | |
PBS with 0.2% BSA, 0.1% gelatin and 0.09% sodium azide; pH 7.2 | |
P07750 | |
Interleukin-4, BSF1, IL-4, Lymphocyte stimulatory factor 1, MGC79402 | |
Rat | |
Affinity chromatography | |
RUO | |
Antibody | |
Monoclonal | |
Murine |
Flow Cytometry | |
BVD6-24G2 | |
FITC | |
Il4 | |
Conjugated | |
Il4 | |
IgG1, kappa | |
25 Tests | |
Store at 2-8°C. Do not freeze. Light-sensitive material. | |
Primary | |
16189 |
For Research Use Only.