missing translation for 'onlineSavingsMsg'
Learn More

IL-4, Biotin, clone: BVD6-24G2, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 13-7042-81

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15310020

  • £62.18 / 50µg


EU Stock 3
Add to basket

Description

Description

Description: The BVD6-24G2 antibody reacts with mouse interleukin-4 (IL-4), a 14kDa cytokine secreted by Th2 cells.Applications Reported: The BVD6-24G2 antibody is used for detection of mouse IL-4 by ELISA.Applications Tested: The BVD6-24G2 antibody has been tested as the detection antibody in a sandwich ELISA for analysis of mouse Interleukin-4 (IL-4) in combination with the purified 11B11 (14-7041) antibody for capture and recombinant mouse IL-4 (14-8041) as the standard. A suitable range of concentrations of this antibody for ELISA detection is 0.5-2 μg/mL. A standard curve consisting of doubling dilutions of the recombinant standard over the range of 500 pg/mL - 4 pg/mL should be included in each ELISA plate.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

IL-4
Monoclonal
0.5mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
P07750
Interleukin-4, BSF1, IL-4, Lymphocyte stimulatory factor 1, MGC79402
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Murine
ELISA
BVD6-24G2
Biotin
Il4
Conjugated
Il4
IgG1, kappa
50μg
Store at 2-8°C. Do not freeze.
Primary
16189
Videos

For Research Use Only.