missing translation for 'onlineSavingsMsg'
Learn More

IL-4, eFluor 660, clone: OX81, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 50-7045-80

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15361940

  • £68.96 / 25µg


EU Stock 3
Add to basket

Description

Description

Description: This OX81 monoclonal antibody reacts with rat interleukin (IL)-4. Also known as BCDF, BCGF-1 and BSF-1, IL-4 is secreted by activated Th2 and natural killer T cells. This cytokine is also produced by Th1 and mast cells albeit to a lesser extent. IL-4 is species-specific and promotes proliferation and differentiation of B cells. IL-4, in conjunction with other signals such as CD40, induces Ig isotype switching of B cells and the upregulation of MHC class II and CD23.The OX81 antibody has been reported to exhibit neutralizing activity.Applications Reported: This OX81 antibody has been reported for use in intracellular staining followed by flow cytometric analysis.Applications Tested: This OX81 antibody has been tested by intracellular flow cytometric analysis of activated rat splenocytes. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.eFluor® 660 is a replacement for Alexa Fluor® 647. eFluor® 660 emits at 659 nm and is excited with the red laser (633 nm). Please make sure that your instrument is capable of detecting this fluorochome.Excitation: 633-647 nm; Emission: 668 nm; Laser: Red Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

IL-4
Monoclonal
0.2mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
P20096
Interleukin-4, BSF1, IL-4, Lymphocyte stimulatory factor 1, MGC79402
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Rat
Flow CytometryþFlow Cytometry
OX81
eFluor 660
Il4
Conjugated
Il4
IgG1, kappa
25μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
287287
Videos

For Research Use Only.