missing translation for 'onlineSavingsMsg'
Learn More

IL-4, APC, clone: 8D4-8, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 17-7049-81

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15300830

  • £102.54 / 50µg


EU Stock 8
Add to basket

Description

Description

Description: The 8D4-8 antibody reacts with human interleukin-4 (IL-4), a 15-19kDa cytokine secreted by Th2 cells.Applications Reported: The 8D4-8 antibody has been reported for use as a capture antibody for human IL-4 ELISA and for intracellular staining for flow cytometric analysis.Applications Tested: This 8D4-8 antibody has been tested in intracellular staining and flow cytometric analysis of stimulated human peripheral blood cells. This can be used at less than or equal to 0.125μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

IL-4
Monoclonal
0.2mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
P05112
Interleukin-4, BSF1, IL-4, Lymphocyte stimulatory factor 1, MGC79402
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow CytometryþFlow Cytometry
8D4-8
APC
IL4
Conjugated
IL4
IgG1, kappa
50μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
3565
Videos

For Research Use Only.