missing translation for 'onlineSavingsMsg'
Learn More

IL-4 Mouse anti-Human, Alexa Fluor® 488, Clone: 8D4-8, eBioscience™

Mouse Monoclonal Antibody

£80.00 - £168.00

Specifications

Antigen IL-4
Clone 8D4-8
Target Species Human
Conjugate Alexa Fluor™ 488
Isotype IgG1, kappa
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15580237
View Documents
Affymetrix eBioscience
53-7049-41
25 Tests £80.00
25 tests View alternative products
Save Up to 
»
Estimated Shipment
03-11-2020
Please to purchase this item. Need a web account? Register with us today!
15590237
missing translation for 'viewDocuments'
Affymetrix eBioscience
53-7049-42
100 Tests £168.00
100 tests Alternative Produkte anzeigen
missing translation for 'toSaveUpto' 
»
Estimated Shipment
03-11-2020
Please to purchase this item. Need a web account? Register with us today!
Beschreibung

Beschreibung

Interleukin-4 (IL-4) is a pleiotropic, immune-modulatory cytokine produced by activated T cells, and serves as a ligand for interleukin 4 receptor. IL-4 has been found to have the broadest range of actions. The interleukin 4 receptor also binds to IL13, which may contribute to many overlapping functions of this cytokine and IL13. STAT6, a signal transducer and activator of transcription, has been shown to play an important role in mediating the immune regulatory signal of IL-4. The IL-4 gene, IL-3, IL-5, IL-13, and CSF2 form a cytokine gene cluster on chromosome 5q. In particular, IL-4 is located in close proximity to IL13 on the chromosome. Further, the IL-4 gene, IL13 and IL5 are found to be coordinated by several long-range regulatory elements over a 120 kilobase range on the chromosome. IL-4 delta 2 is a known splice variant of IL-4.
Spezifikation

Spezifikation

IL-4
Human
IgG1, kappa
Monoclonal
Liquid
Antibody
4° C, store in dark, DO NOT FREEZE!
8D4-8
Alexa Fluor™ 488
Flow Cytometry
Primary
Monoclonal
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
RUO

For Research Use Only.