Learn More
IL-38 Monoclonal Antibody (H127C), eFluor™ 660, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
£97.00 - £248.00
Specifications
Antigen | IL-38 |
---|---|
Clone | H127C |
Classification | Monoclonal |
Conjugate | eFluor 660 |
Form | Liquid |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15510047
|
Affymetrix eBioscience
50-7385-82 |
100 μg |
£248.00
100µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15500047
|
Affymetrix eBioscience
50-7385-80 |
25 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Description: The monoclonal antibody H127C recognizes human IL-38. IL-38 is a member of the Interleukin-1 family (IL-1F) and shares homology with IL-1R antagonist (IL-1Ra) and IL-36R antagonist (IL-36Ra). IL-38 is expressed mostly in the skin and proliferating B cells of the tonsil and is also present at low levels in the heart and placenta. IL-38 functions as an anti-inflammatory cytokine that inhibits IL-36 by binding to IL-36R and also inhibits the production of IL-17 and IL-22. Gene polymorphisms of IL-38 are associated with psoriatic arthritis, ankylosing spondylitis, and heart disease. Applications Reported: This H127C antibody has been reported for use in immunohistochemical staining of formalin-fixed paraffin embedded tissue sections. Applications Tested: This H127C antibody has been tested tested by immunohistochemistry on human FFPE tissue using low pH antigen retrieval and can be used at less than or equal to 5 μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. eFluor™ 660 is a replacement for Alexa Fluor™ 647. eFluor™ 660 emits at 659 nm and is excited with the red laser (633 nm). Please make sure that your instrument is capable of detecting this fluorochome. Excitation: 633-647 nm; Emission: 668 nm; Laser: Red Laser. Filtration: 0.2 μm post-manufacturing filtered.
IL-38 Monoclonal antibody specifically detects IL-38 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).Specifications
IL-38 | |
Monoclonal | |
Liquid | |
RUO | |
Q8WWZ1 | |
IL1F10 | |
Primary | |
Antibody |
H127C | |
eFluor 660 | |
Mouse | |
Human | |
84639 | |
IgG2b κ | |
Affinity chromatography | |
IL1F10 |
For Research Use Only.