missing translation for 'onlineSavingsMsg'
Learn More

IL-33R (ST2), PerCP-eFluor™ 710, clone: RMST2-33, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 46-9333-82

Code : NEW

Additional Details : Weight : 0.09500kg

 View more versions of this product

Product Code. 15519416

  • £205.21 / 100µg


EU Stock 1
Add to basket

Description

Description

Description: The RMST2-33 monoclonal antibody reacts with mouse ST2, also known as IL-33R or interleukin 1 receptor-like 1. ST2 is a member of the IL-1 receptor family. In the membrane-bound form, it consists of three extracellular immunoglobulin domains and an intracellular toll-interleukin-1 receptor domain. ST2 forms a dimer with IL-1R accessory protein in a ligand-dependent manner. IL-33/ST2 interaction induces the production of TH2 cytokines. ST2 also has a soluble isoform lacking transmembrane and intracellular toll-interleukin-1 receptor domains. It is believed that the soluble form functions as a decoy receptor that can block membrane bound IL-33/ST2 interaction. ST2 is expressed by TH2 lymphocytes, mast cells, eosinophils, basophils, innate lymphocytes, smooth muscle cells, and endothelial cells and is involved in host defense, allergy, and inflammation. IL-33/ST2 interaction has also been shown to be atheroprotective.RMST2-33 has been reported to bind and activate ST2 in in vitro assays.Applications Reported: This RMST2-33 antibody has been reported for use in flow cytometric analysis.Applications Tested: This RMST2-33 antibody has been tested by flow cytometric analysis of mouse bone marrow-derived mast cells. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.PerCP-eFluor® 710 emits at 710 nm and is excited with the blue laser (488 nm); it can be used in place of PerCP-Cyanine5.5. We recommend using a 710/50 bandpass filter, however, the 695/40 bandpass filter is an acceptable alternative. Please make sure that your instrument is capable of detecting this fluorochrome.Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light.Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 μL of cell sample + 100 μL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically.Excitation: 488 nm; Emission: 710 nm; Laser: Blue Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

IL-33R (ST2)
Monoclonal
0.2mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
P14719
DER4, Fit-1, Ly84, ST2L, St2, St2-rs1, T1, T1/ST2
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Murine
Flow Cytometry
RMST2-33
PerCP-eFluor 710
Il1rl1
Conjugated
Il1rl1
IgG2b, kappa
100μg
Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage, handling and experimental procedures.
Primary
17082
Videos

For Research Use Only.