missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-31 Rat anti-Mouse, Clone: 323407, R&D Systems™
Rat Monoclonal Antibody
POA
Specifications
Antigen | IL-31 |
---|---|
Clone | 323407 |
Applications | Western Blot |
Conjugate | Unconjugated |
Form | Purified |
Description
IL-31 Monoclonal specifically detects IL-31 in Mouse samples. It is validated for Western Blot.Specifications
IL-31 | |
Western Blot | |
Purified | |
RUO | |
386653 | |
Monoclonal | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
323407 | |
Unconjugated | |
Rat | |
Mouse | |
E. coli-derived recombinant mouse IL-31 Thr24-Cys163 Accession # Q6EAL8 | |
Primary |
For Research Use Only