missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-31 Mouse anti-Human, eFluor® 660, Clone: 31SNEZE, eBioscience™
Mouse Monoclonal Antibody
Brand: Affymetrix eBioscience 50-9319-41
Code : NEW
Additional Details : Weight : 0.23750kg
Description
eFluor® 660 is a replacement for Alexa Fluor® 647. eFluor® 660 emits at 659 nm and is excited with the red laser (633 nm). Please make sure that your instrument is capable of detecting this fluorochome. Excitation: 633-647 nm; Emission: 668 nm; Laser: Red Laser. Filtration: 0.2 μm post-manufacturing filtered. Interleukin 31, or IL-31 is a cytokine secreted by differentiated CD4+ T helper type 2 (Th2) cells. It is also released during inflammation of the skin, by macrophages and T cells. Because IL-31 is secreted by activated Th2 cells, it has a role in host defense against infection and may be involved in asthma, allergy and different types of bowel diseases.Specifications
IL-31 | |
Monoclonal | |
5 μL/Test | |
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2 | |
Q6EBC2 | |
Interleukin-31 | |
Mouse | |
Affinity chromatography | |
RUO | |
Antibody | |
Monoclonal | |
Human |
Flow Cytometry | |
31SNEZE | |
eFluor™ 660 | |
IL31 | |
Liquid | |
IL31 | |
IgG2b, kappa | |
25 Tests | |
4° C, store in dark, DO NOT FREEZE! | |
Primary | |
386653 |
For Research Use Only.