missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-3R beta Rat anti-Mouse, Clone: 130710, R&D Systems™
Rat Monoclonal Antibody
POA
Specifications
Antigen | IL-3R beta |
---|---|
Clone | 130710 |
Immunogen | Mouse myeloma cell line NS0-derived recombinant mouse IL-3 R beta His23-Trp440 Accession # P26954 |
Gene ID (Entrez) | 1439 |
Gene Accession No. | P26954 |
Description
IL-3R beta Monoclonal specifically detects IL-3R beta in Mouse samples. It is validated for ELISA Capture (Matched Antibody Pair).Specifications
IL-3R beta | |
Mouse myeloma cell line NS0-derived recombinant mouse IL-3 R beta His23-Trp440 Accession # P26954 | |
P26954 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Rat | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
RUO |
130710 | |
1439 | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Primary | |
Detects mouse IL-3 R beta in ELISAs. In sandwich immunoassays, no cross-reactivity or interference with recombinant mouse IL-3 R alpha or recombinant human IL-3 R alpha is observed. Cross-reactivity with AIC2B was not tested. | |
Unconjugated | |
ELISA | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only