missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-3R alpha/CD123 Goat anti-Mouse, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
POA
Specifications
Antigen | IL-3R alpha/CD123 |
---|---|
Immunogen | S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse IL-3 R alpha/CD123 Ser17-Lys331 Accession # P26952 |
Gene ID (Entrez) | 3563 |
Gene Accession No. | P26952 |
Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative |
Description
IL-3R alpha/CD123 Polyclonal specifically detects IL-3R alpha/CD123 in Mouse samples. It is validated for Western Blot.Specifications
IL-3R alpha/CD123 | |
3563 | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Primary | |
Detects mouse IL-3 R alpha/CD123 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant human IL-3 R alpha and recombinant mouse IL-3 R beta is observed. | |
Unconjugated | |
Western Blot | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse IL-3 R alpha/CD123 Ser17-Lys331 Accession # P26952 | |
P26952 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Goat | |
IgG | |
Affinity Purified | |
RUO |
For Research Use Only