missing translation for 'onlineSavingsMsg'
Learn More

IL-28 Rabbit anti-Mouse, Biotin, Polyclonal, eBioscience™

Rabbit Polyclonal Antibody

Brand:  Affymetrix eBioscience 13-7282-85

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15360050

  • £170.59 / 500µg


Estimated Shipment date
from Supplier 29-06-2018
Add to basket

Description

Description

This polyconal Anti-Mouse IL-28 antibody reacts with mouse interleukin-28 (IL-28), a member of the IFN-lambda family which mediates the induction of anti-viral protection in a wide variety of cells.
Specifications

Specifications

IL-28
0.5mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Interleukin-28
IgG
RUO
Primary
Murine
ELISA
Biotin
Conjugated
Rabbit
500μg
Store at 2-8°C. Do not freeze.
Polyclonal
Videos

For Research Use Only.