missing translation for 'onlineSavingsMsg'
Learn More

EBI3 (IL-27 subunit), APC, clone: ebic6, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 17-7358-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15558196

  • £81.61 / 25 tests


Estimated Shipment date
from Supplier 23-02-2018
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: This ebic6 monoclonal antibody reacts with human Epstein-Barr Virus Induced Gene 3 (EBI3). EBI3 can be expressed and secreted as monomers or homodimers, and was first identified in virally-infected B cells. It is also believed to be secreted as a homodimer by cells of the trophoblast, where it may play a role in immune tolerance during pregnancy.Additionally, EBI3 is a component of two of the four members of the interleukin (IL)-12 family of heterodimeric cytokines. EBI3 was first found to heterodimerize with p28 (IL-30) to form IL-27, which is secreted by antigen-presenting cells in response to pro-inflammatory stimuli. IL-27 has been shown to have both pro-inflammatory and anti-inflammatory effects. It influences the commitment of CD4+ T-cells toward the Th1 lineage by inducing the expression of the T-bet transcription factor and the upregulation of IL-12R beta 2. Its anti-inflammatory functions include the suppression of Th2 and Th17 proliferation and differentiation.More recently, EBI3 was also found to dimerize with the IL-12 subunit, p35 to form IL-35. The biology of IL-35 is incompletely understood, but it is believed to be unique among the IL-12 family members in its expression and function. It is produced by regulatory T cells, and appears to have immunoregulatory functions. IL-35 has been found to suppress the proliferation and function of effector T cells, and has also been observed to induce a regulatory phenotype on naive CD4+ cells cultured in its presence.ELISA results from the culture of human dendritic cells stimulated shown above suggests that at least some of the EBI3 detected by flow is dimerized with p28 to form IL-27. Testing with ebic6 in the context of IL-35 is currently in process. For more information, please contact technical support.Applications Reported: This ebic6 antibody has been reported for use in intracellular staining followed by flow cytometric analysis.Applications Tested: This ebic6 antibody has been pre-titrated and tested by intracellular staining and flow cytometric analysis of stimulated normal human peripheral blood cells using the Intracellular Fixation & Permeabilization Buffer Set (cat. 88-8824) and protocol. Please refer to Best Protocols: Protocol A: Two step protocol for (cytoplasmic) intracellular proteins located under the Resources Tab online. This antibody can be used at 5 μL (0.06 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

IL-27 EBI3
ebic6
APC
Conjugated
Murine
25 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Flow Cytometry (Intracellular Staining)
5μL (0.06μg)/test
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Interleukin-27, Epstein-Barr Virus-Induced gene 3
IgG2b
RUO
Primary
Human
Videos