missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-23 p19, Functional Grade, clone: HNU2319, eBioscience™
Mouse Monoclonal Antibody
Brand: Affymetrix eBioscience 16-5236-85
Code : Z2
Additional Details : Weight : 0.01000kg
Description
Description: The HNU2319 monoclonal antibody is specific to the p19 subunit of human IL-23, a heterodimeric cytokine made up of two covalently linked subunits, p40 and p19. It is closely related to IL-12, with which it shares the p40 subunit. Dendritic cells and macrophages produce IL-23 in response to TLR2, TLR4, and TLR8 ligands, as well as the beta-glucan receptor Dectin-1. Human IL-23 induces proliferation of memory T cells and induces moderate levels of IFN-gamma production by naive and memory T cells, as compared to IL-12. In contrast, the mouse IL-23 biological activities are more distinctly different than those of mouse IL-12 suggesting the signaling or response elements (receptors) differ between species. The IL-23 receptor is also heterodimeric and shares the IL-12Rbeta1 chain with IL-12, while the IL-23R chain is unique to IL-23. Signaling occurs through the Jak/STAT pathway and results in RORgammat expression, which drives the differentiation of CD4^+ T-cells towards the Th17 phenotype. The monoclonal antibody HNU2319 neutralizes the bioactivity of IL-23 without affecting IL-12.Applications Reported: The monoclonal antibody HNU2319 reacts with the p19 subunit of human IL-23 and inhibits its bioactivity.Applications Tested: The ND50 of HNU2319, as determined by the neutralization of mouse IL-17A induction in balb/c splenocytes by recombinant human IL-23, is 0.2-0.4 μg/mL in the presence of 2 ng/mL IL-23. This antibody is specific to the p19 subunit of IL-23, and will not affect the bioactivity of IL-12. The neutralization dose of this antibody will vary depending on cell type, assay method, and concentration of cytokine.Endotoxin: Less than 0.001 ng/ug antibody as determined by the LAL assay.Storage and handling: Use in a sterile environment.Filtration: 0.2μm post-manufacturing filtered.Purity: Greater than 90%, as determined by SDS-PAGE.Endotoxin Level: Less than 0.001 ng/μg antibody, as determined by LAL assay.Aggregation: Less than 10%, as determined by HPLC.Specifications
IL-23 p19 | |
Monoclonal | |
1mg/mL | |
PBS with no preservative; pH 7.2 | |
Q9NPF7 | |
IL-23, IL-23A, IL23P19, MGC79388, P19, SGRF | |
Murine | |
Affinity chromatography | |
RUO | |
Antibody | |
Monoclonal | |
Human |
Cytokine Neutralization | |
HNU2319 | |
Functional Grade | |
IL23A | |
Purified | |
IL23A | |
IgG1, kappa | |
500μg | |
Store at 2-8°C. | |
Primary | |
51561 |
For Research Use Only.