missing translation for 'onlineSavingsMsg'
Learn More

IL-21, PE, clone: mhalx21, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 12-7213-82

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15269309

  • £205.37 / 100µg


EU Stock 1
Add to basket

Description

Description

Description: The monoclonal antibody mhalx21 reacts with mouse interleukin-21 (IL-21). IL-21 is a 17kDa immunomodulatory cytokine produced mainly by NKT, T helper (Th) 17 and T follicular helper (TFH) cells. In TFH cells, IL-21 expression leads to autocrine signaling through the IL-21 receptor (IL-21R) and STAT3, which leads to additional transcriptional activation by Bcl6. As with IFNgamma for Th1, IL-4 for Th2 and IL-17A for Th17, IL-21 is critical for TFH cell effector function. This cytokine plays a role in T cell-dependent B cell differentiation into plasma cells and memory cells, stimulation of IgG production and induction of apoptotic signaling in naive B cells. In Th17 cells, IL-21 expression and autocrine feedback through STAT3, IRF4 and RORgammat lead to upregulation of the IL-23R, thereby preparing Th17 cells for maturation and maintenance by the inflammatory cytokine IL-23. While upregulating IRF4 and RORgammat, IL-21 also mediates the downregulation of Foxp3. High levels of IL-21 are present in chemically-induced colitis models. IL-21-deficient mice are protected from developing colitis upon chemical treatment by their inability to upregulate Th17-associated molecules.Preliminary data suggest that clone mhalx21 recognizes a different epitope than anti-mouse IL-21 clone FFA21 (cat. 51-7211).Applications Reported: This mhalx21 antibody has been reported for use in intracellular staining followed by flow cytometric analysis. This clone has been optimized for use with eBioscience IC Fixation and Permeabilization kit (cat. 88-8823).Applications Tested: This mhalx21 antibody has been tested on Th17-polarized splenocytes restimulated with PMA, Ionomycin and Brefeldin A for 5 hours. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

IL-21
Monoclonal
0.2mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
Q9ES17
Interleukin-21, IL-21, Za11
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Murine
Flow Cytometry
mhalx21
PE
Il21
Conjugated
Il21
IgG2a, kappa
100μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
60505
Videos

For Research Use Only.