Learn More
IL-21 Monoclonal Antibody (FFA21), APC, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
Brand: Affymetrix eBioscience 17-7211-80
Code : Z2
Additional Details : Weight : 0.25000kg
Description
Description: This FFA21 monoclonal antibody reacts with mouse interleukin-21 (IL-21). IL-21 is a 17 kDa immunomodulatory cytokine produced mainly by NKT, T helper (Th) 17 and T follicular helper (TFH) cells. In TFH cells, IL-21 expression leads to autocrine signaling through the IL-21 receptor (IL-21R) and STAT3, which leads to additional transcriptional activation by Bcl6. As with IFN gamma for Th1, IL-4 for Th2 and IL-17A for Th17, IL-21 is critical for TFH cell effector function. This cytokine plays a role in T cell-dependent B cell differentiation into plasma cells and memory cells, stimulation of IgG production and induction of apoptotic signaling in naive B cells. In Th17 cells, IL-21 expression and autocrine feedback through STAT3, IRF4 and ROR gamma t lead to upregulation of the IL-23R, thereby preparing Th17 cells for maturation and maintece by the inflammatory cytokine IL-23. While upregulating IRF4 and ROR gamma t, IL-21 also mediates the downregulation of Foxp3. High levels of IL-21 are present in chemically-induced colitis models. IL-21-deficient mice are protected from developing colitis upon chemical treatment by their inability to upregulate Th17-associated molecules. Preliminary data suggest that clone FFA21 recognizes a different epitope than anti-mouse IL-21 clone mhalx21 (cat. 51-7213). Applications Reported: This FFA21 antibody has been reported for use in intracellular staining followed by flow cytometric analysis.
IL-21 Monoclonal antibody specifically detects IL-21 in Mouse samples. It is validated for Flow Cytometry.Specifications
IL-21 | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
Q9ES17 | |
IL21 | |
Affinity chromatography | |
RUO | |
60505 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
FFA21 | |
APC | |
IL21 | |
CVID11; CX2; il 21; IL21; IL-21; ILN; Interleukin; interleukin 21; Interleukin21; interleukin-21; interleukin-21 isoform; porcine interleukin-21; Za11 | |
Rat | |
25 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2a κ |
For Research Use Only.