Learn More
IL-21 Monoclonal Antibody (eBio3A3-N2 (3A3-N2)), eFluor™ 660, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Affymetrix eBioscience 50-7219-41
Code : NEW
Additional Details : Weight : 0.23750kg
Description
Description: The monoclonal antibody eBio3A3-N2 reacts with human IL-21, a potent stimulator of NK and cytolytic T cells. IL-21 is a 131 amino acid protein most closely related to IL-2 and IL-15. Expression is found in CD4+ cells upon activation. In addition to its effect on NK cells, IL-21 stimulates proliferation of B-cell stimulated by crosslinking of the CD40 antigen and stimulates the proliferation of bone marrow progenitor cells and the expression of the NK-cell marker CD56 in the presence of IL-15. Applications Reported: This eBio3A3-N2 (3A3-N2) antibody has been reported for use in intracellular staining followed by flow cytometric analysis. Applications Tested: This eBio3A3-N2 (3A3-N2) antibody has been pre-titrated and tested by intracellular staining and flow cytometric analysis of restimulated, Th17-polarized normal human peripheral blood cells. This can be used at 5 μL (0.125 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Staining for IL-21 has shown donor variability. stimulation times may vary depending on the donor and should be optimized. Increased staining levels have been observed on Th17-polarized normal human peripheral blood cells. eFluor™ 660 is a replacement for Alexa Fluor™ 647.
IL-21 Monoclonal antibody specifically detects IL-21 in Human samples. It is validated for Flow Cytometry.Specifications
IL-21 | |
Monoclonal | |
5 μL/Test | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2 | |
Q9HBE4 | |
IL21 | |
Affinity chromatography | |
RUO | |
59067 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
eBio3A3-N2 (3A3-N2) | |
eFluor 660 | |
IL21 | |
CVID11; CX2; il 21; IL21; IL-21; ILN; Interleukin; interleukin 21; Interleukin21; interleukin-21; interleukin-21 isoform; porcine interleukin-21; Za11 | |
Mouse | |
25 Tests | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
For Research Use Only.