Learn More
IL-21, clone: eBio3A3-N2 (3A3-N2), eBioscience™
Mouse Monoclonal Antibody
Brand:  Affymetrix eBioscience 14-7219-80
Code : Z2
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15218047
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »
Disclaimers:
For Research Use Only.
Description
Description
The monoclonal antibody eBio3A3-N2 reacts with human IL-21, a potent stimulator of NK and cytolytic T cells. IL-21 is a 131 amino acid protein most closely related to IL-2 and IL-15. Expression is found in CD4+ cells upon activation. In addition to its effect on NK cells, IL-21 stimulates proliferation of B-cell stimulated by crosslinking of the CD40 antigen and stimulates the proliferation of bone marrow progenitor cells and the expression of the NK-cell marker CD56 in the presence of IL-15.
Specifications
Specifications
IL-21
eBio3A3-N2 (3A3-N2)
Unconjugated
Purified
Murine
25μg
Store at 2-8°C.
Monoclonal
ELISA Capture
0.5mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Interleukin-21, IL21
IgG1