missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-20 R beta Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 2 publications
POA
Specifications
Antigen | IL-20 R beta/FNDC6 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
IL-20 R beta/FNDC6 Polyclonal specifically detects IL-20 R beta/FNDC6 in Human samples. It is validated for Western Blot.Specifications
IL-20 R beta/FNDC6 | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Goat | |
Human | |
FNDC6, DIRS1, FNDC6, IL20R beta, IL-20R2, IL-20Rb, IL20RB, IL20RB interleukin 20 receptor beta, MGC34923 | |
IL20RB | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
Q6UXL0 | |
53833 | |
Mouse myeloma cell line NS0-derived recombinant human IL-20 R beta Asp30-Ala230 Accession # Q6UXL0 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human IL-20 R beta in direct ELISAs and Western blots. In direct ELISAs, approximately 60% cross-reactivity with recombinant mouse IL-20 R beta is observed and less than 2% cross-reactivity with recombinant human (rh) IL-20 R alpha , rhIL-10 R beta , rhIFN-gamma R1, and rhIFN-gamma R2 is observed. |
For Research Use Only