Learn More
IL-2 Monoclonal Antibody (JES6-5H4), PerCP-Cyanine5.5, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
£89.75 - £252.00
Specifications
Antigen | IL-2 |
---|---|
Clone | JES6-5H4 |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15301210
|
Affymetrix eBioscience
45-7021-80 |
25 μg |
£89.75
25µg |
Estimated Shipment: 28-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15311210
|
Affymetrix eBioscience
45-7021-82 |
100 μg |
£252.00
100µg |
Estimated Shipment: 29-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Description: The JES6-5H4 antibody reacts with mouse interleukin-2 (IL-2), a 17 kDa T cell growth factor and a major immunoregulatory cytokine. Applications Reported: This JES6-5H4 antibody has been reported for use in intracellular staining followed by flow cytometric analysis. Applications Tested: This JES6-5H4 antibody has been tested by intracellular staining and flow cytometric analysis of stimulated mouse splenocytes. This can be used at less than or equal to 0.125 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 488 nm; Emission: 695 nm; Laser: Blue Laser. Filtration: 0.2 μm post-manufacturing filtered.
IL-2 Monoclonal antibody specifically detects IL-2 in Mouse samples. It is validated for Flow Cytometry.Specifications
IL-2 | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
aldesleukin; cytokine; H-IL-2; il 2; Il2; Il-2; ILN; Interleukin; interleukin 2; interleukin 2 precursor; interleukin 2 variant IL2delta2; Interleukin2; Interleukin-2; interleukin-2 precursor; interleukin-2 precursor protein; interleukin-2; interleukin 2; Involved in regulation of T cell clonal expansion; involved in regulation of T-cell clonal expansion; lymphokine; M-IL-2; patial sequence of Interleukin-2; POIL2; porcine Interleukin 2; T cell growth factor; T-cell growth factor; T-cell growth factor (TCGF); TCGF | |
IL2 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
IL2 |
JES6-5H4 | |
Flow Cytometry | |
PerCP-Cyanine5.5 | |
Rat | |
Mouse | |
P04351 | |
16183 | |
IgG2b κ | |
Affinity chromatography | |
Antibody |
For Research Use Only.