missing translation for 'onlineSavingsMsg'
Learn More

IL-2 Rat anti-Mouse, PE-Cyanine7, Clone: JES6-5H4, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 25-7021-82

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15341000

  • £144.00 / 100µg


EU Stock 4
Add to basket

Description

Description

Filtration: 0.2 μm post-manufacturing filtered. Interleukin 2 (IL-2) is an immuno-modulatory cytokine that is important for the proliferation of activated T cells, differentiation of B cells, natural killer cells, monocytes and macrophages. IL-2 signals through the IL-2 receptor (IL-2R), a heterotrimeric protein complex whose gamma chain is also shared by interleukin 4 (IL-4) and interleukin 7 (IL-7). The expression of the IL-2 gene in mature thymocytes is monoallelic, which represents an unusual regulatory mode for controlling the precise expression of a single gene. The targeted disruption of a gene similar to IL-2 in mice leads to an ulcerative colitis-like disease that suggests an essential role of this gene in the immune response to antigenic stimuli.
Specifications

Specifications

IL-2
Monoclonal
0.2 mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
P04351
Interleukin-2, T-cell growth factor, TCGF, Aldesleukin
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Mouse
Flow Cytometry
JES6-5H4
PE-Cyanine7
Il2
Liquid
Il2
IgG2b, kappa
100 μg
4° C, store in dark, DO NOT FREEZE!
Primary
16183
Videos

For Research Use Only.