missing translation for 'onlineSavingsMsg'
Learn More

IL-2, FITC, clone: JES6-5H4, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 11-7021-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15536586

  • £56.31 / 25 tests


Estimated Shipment date
from Supplier 26-03-2018
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: The JES6-5H4 antibody reacts with mouse interleukin-2 (IL-2), a 17kDa T cell growth factor and a major immunoregulatory cytokine.Applications Reported: JES6-5H4 has been reported for use in intracellular staining and flow cytometric analysis.Applications Tested: The JES6-5H4 has been pre-titrated and tested by intracellular staining and flow cytometric analysis. Refer to catalog number suffix on the vial for amount to use per test: -41 is 5μL (0.25μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

IL-2
JES6-5H4
FITC
Conjugated
Rat
25 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Flow Cytometry (Intracellular Staining)
0.5mg/mL;
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Interleukin-2
IgG2b
RUO
Primary
Murine
Videos