Learn More
IL-2 Monoclonal Antibody (JES6-5H4), eFluor™ 450, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
£82.75 - £208.00
Specifications
Antigen | IL-2 |
---|---|
Clone | JES6-5H4 |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15381820
|
Affymetrix eBioscience
48-7021-82 |
100 μg |
£208.00
100µg |
Estimated Shipment: 16-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15371820
|
Affymetrix eBioscience
48-7021-80 |
25 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Description: The JES6-5H4 antibody reacts with mouse interleukin-2 (IL-2), a 17 kDa T cell growth factor and a major immunoregulatory cytokine. Applications Reported: This JES6-5H4 antibody has been reported for use in intracellular staining followed by flow cytometric analysis. Applications Tested: This JES6-5H4 antibody has been tested by intracellular staining and flow cytrometric analysis of stimulated mouse splenocytes. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. eFluor™ 450 is an alternative to Pacific Blue™. eFluor™ 450 emits at 445 nm and is excited with the Violet laser (405 nm). Please make sure that your instrument is capable of detecting this fluorochome. Excitation: 405 nm; Emission: 445 nm; Laser: Violet Laser. Filtration: 0.2 μm post-manufacturing filtered.
IL-2 Monoclonal antibody specifically detects IL-2 in Mouse samples. It is validated for Flow Cytometry.Specifications
IL-2 | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
P04351 | |
IgG2b κ | |
Affinity chromatography | |
Antibody |
JES6-5H4 | |
Flow Cytometry | |
eFluor 450 | |
Rat | |
Mouse | |
16183 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! |
For Research Use Only.