Learn More
IL-2 Monoclonal Antibody (JES6-5H4), eBioscience™, Invitrogen™
Rat Monoclonal Antibody
£65.75 - £191.00
Specifications
Antigen | IL-2 |
---|---|
Clone | JES6-5H4 |
Concentration | 0.5 mg/mL |
Applications | Flow Cytometry, Immunoprecipitation |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15297977
|
Affymetrix eBioscience
14-7021-81 |
50 μg |
£65.75
50µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15207987
|
Affymetrix eBioscience
14-7021-85 |
500 μg |
£191.00
500µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Description: The JES6-5H4 antibody reacts with mouse interleukin-2 (IL-2), a 17 kDa T cell growth factor and a major immunoregulatory cytokine. Applications Reported: The purified JES6-5H4 has been reported for use in blocking of fluorochrome conjugated JES6-5H4 in intracellular flow cytometry experiments to serve as specificity control. JES6-5H4 is reported for detection of mouse IL-2 by ELISA, immunoprecipitation, and for neutralizing IL-2 binding and biological activity (use Functional Grade purified). Applications Tested: The JES6-5H4 antibody has been tested by sandwich ELISA for detection of mouse IL-2 in combination with JES6-1A12 and can be used between 0.5-2 μg/mL. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered.
Interleukin 2 (IL-2) is an immuno-modulatory cytokine that is important for the proliferation of activated T cells, differentiation of B cells, natural killer cells, monocytes and macrophages. IL-2 signals through the IL-2 receptor (IL-2R), a heterotrimeric protein complex whose gamma chain is also shared by interleukin 4 (IL-4) and interleukin 7 (IL-7). The expression of the IL-2 gene in mature thymocytes is monoallelic, which represents an unusual regulatory mode for controlling the precise expression of a single gene. The targeted disruption of a gene similar to IL-2 in mice leads to an μgcerative colitis-like disease that suggests an essential role of this gene in the immune response to antigenic stimuli.Specifications
IL-2 | |
0.5 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
aldesleukin; cytokine; H-IL-2; il 2; Il2; Il-2; ILN; Interleukin; interleukin 2; interleukin 2 precursor; interleukin 2 variant IL2delta2; Interleukin2; Interleukin-2; interleukin-2 precursor; interleukin-2 precursor protein; interleukin-2; interleukin 2; Involved in regulation of T cell clonal expansion; involved in regulation of T-cell clonal expansion; lymphokine; M-IL-2; patial sequence of Interleukin-2; POIL2; porcine Interleukin 2; T cell growth factor; T-cell growth factor; T-cell growth factor (TCGF); TCGF | |
IL2 | |
Primary | |
4° C | |
IL2 |
JES6-5H4 | |
Flow Cytometry, Immunoprecipitation | |
Unconjugated | |
Rat | |
Mouse | |
P04351 | |
16183 | |
IgG2b κ | |
Affinity chromatography | |
Antibody |
For Research Use Only.