Learn More
IL-2 Monoclonal Antibody (JES6-5H4), Alexa Fluor™ 488, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
Brand: Affymetrix eBioscience 53-7021-82
Code : Z2
Additional Details : Weight : 0.25000kg
Description
Description: The JES6-5H4 antibody reacts with mouse interleukin-2 (IL-2), a 17 kDa T cell growth factor and a major immunoregulatory cytokine. Applications Reported: The JES6-5H4 antibody has been reported useful for ELISA, ELISPOT, neutralization, and intracellular staining for flow cytometric analysis. Applications Tested: The Alexa Fluor™ 488 JES6-5H4 antibody has been tested by intracellular staining and flow cytometric analysis and can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 488 nm; Emission: 519 nm; Laser: Blue Laser. Filtration: 0.2 μm post-manufacturing filtered.
IL-2 Monoclonal antibody specifically detects IL-2 in Mouse samples. It is validated for Flow Cytometry.Specifications
IL-2 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
P04351 | |
IL2 | |
Affinity chromatography | |
RUO | |
16183 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
JES6-5H4 | |
Alexa Fluor 488 | |
IL2 | |
aldesleukin; cytokine; H-IL-2; il 2; Il2; Il-2; ILN; Interleukin; interleukin 2; interleukin 2 precursor; interleukin 2 variant IL2delta2; Interleukin2; Interleukin-2; interleukin-2 precursor; interleukin-2 precursor protein; interleukin-2; interleukin 2; Involved in regulation of T cell clonal expansion; involved in regulation of T-cell clonal expansion; lymphokine; M-IL-2; patial sequence of Interleukin-2; POIL2; porcine Interleukin 2; T cell growth factor; T-cell growth factor; T-cell growth factor (TCGF); TCGF | |
Rat | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2b κ |
For Research Use Only.